Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Timolo 99%

Timolo 99%

Bottiglia di plastica, 100 g
Catalog # ACRO150331000
Irritant
Corrosive
Environment
Timolo 99%
Catalog # ACRO150331000
CAS Number:  89-83-8
Timolo 99%
Timolo 99%

Bottiglia di plastica, 100 g

CAS Number:  89-83-8
Price
SFr. 61.20
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  4-Isopropyl-3-hydroxytoluene, 2-Isopropyl-5-methylphenol, 6-Isopropyl-m-cresol
  • CAS Number:89-83-8
  • Formula:2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
  • MDL Number:MFCD00002309
  • Molecular Weight:150.22 g/mol
  • Merck Index:13,09476
  • Boiling Pt:232 °C (760 mmHg)
  • Melting Pt:48…52 °C
  • Density:0,96 g/cm³
  • Flash Pt:102 °C
  • Storage Temperature:Ambiente
  • EINECS:201-944-8
  • UN:2430
  • ADR:8,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    48 to 52 °C
  • GC:
    ≥98.5 %
  • Residue after evaporation:
    ≤0.05 %

Specifications

  • Tipo di conf.:
    Bottiglia di plastica
  • Conf:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.