Isonicotinic acid 99%
Bottiglia di plastica, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Pyridine-4-carboxylic acid, 4-Pyridinecarboxylic acid, 4-Picolinic acid
- CAS Number:55-22-1
- Formula:C₆H₅NO₂
- MDL Number:MFCD00006429
- Molecular Weight:123.11 g/mol
- Merck Index:13,05206
- Boiling Pt:260 °C (15 mmHg)
- Melting Pt:310…315 °C
- Storage Temperature:Ambiente
- EINECS:200-228-2
Documents
Specifications
Specification Test Results
- Appearance:White to cream Powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥98.5 %
- HPLC:≥97.5 %
- Specific optical rotation:-190° to -204° (20°C, 589 nm) (c=2,acetone)
Specifications
- Tipo di conf.:Bottiglia di plastica
- Conf:500 g



