Citidina 99+%
Bottiglia di vetro, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one
- CAS Number:65-46-3
- Formula:C₉H₁₃N₃O₅
- MDL Number:MFCD00006545
- Molecular Weight:243.22 g/mol
- Melting Pt:210…217 °C (dec.)
- Storage Temperature:Refrigerata
- EINECS:200-610-9
Documents
Specifications
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:210 to 217 °C (Dec)
- Loss on drying:≤0.5 % (105 °C, 5 hrs)
- Sulfated ash:≤0.1 % (800 °C)
- HPLC:≥99.0 %
- UV spectrophotometry:98 to 102 % (lambda max. 280 nm, pH 1)
- UV transmittance:≥98 % (c=1, H2O, 430 nm) 1 cm cell
- Ratio of Absorbances:0.43 to 0.47 (A250/A260)
- Heavy metals (as Pb):≤10 ppm
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



