L(-)-Glutatione (forma ridotta), Millipore®
Glass bottle for solids, 5 g
3541-5
: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
- :70-18-8
- :C₁₀H₁₇N₃O₆S
- :MFCD00065939
- :307.33 g/mol
- :13,04488
- :185…195 °C
- :1,475 g/cm³ (20 °C)
- :Refrigerata
- :200-725-4
- Tipo di conf.:Glass bottle for solids
- Conf:5 g