Hydrometers, Brix
0 - 30 °Bx, 0,5 °Bx, 300 mm
Catalog # ALLA3910FC000/20QP
Supplier: Alla France
Jump to:
Product Details & Documents
These Brix sugar scale hydrometers are specially designed for use in breweries.
Specifications
- Lunghezza:300 mm
- Divisione:0,5 °Bx
- Range di misura:0 - 30 °Bx