Soil hydrometers
Soil hydrometer, 0,995 − 1,038 S.G., 0,001 S.G., 280 mm
Catalog # 614-1534
Supplier: Alla France
Jump to:
Product Details & Documents
Soil hydrometers for specific gravity (SG) measurements.
- Medium surface tension (55 mN/m)
- Reading below meniscus
Certifications: ASTM E100 D422 - A2LA traceable.
Ordering information: EU traceable calibration certificate or EO official calibration certificate available on request.
Specifications
- Descrizione:Soil hydrometer
- Lunghezza:280 mm
- Seq:01
- Divisione:0,001 S.G.
- Range di misura:0,995 − 1,038 S.G.