Cupferron
Bottiglia di plastica, 500 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: Ammonium 2-oxo-1-phenylhydrazinolate
- CAS Nummer:135-20-6
- Formel:C₆H₉N₃O₂
- MDL-Nummer:MFCD00078422
- Molekulargewicht:155.16 g/mol
- Schmelzender Platingehalt:150…155 °C (dec.)
- Lagertemperatur:Refrigerata
- EINECS:205-183-2
- UN:2811
- ADR:6.1,III
Documents
ACRO42360
Scheda di sicurezza
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White to yellow-brown Crystalline powder or flakes
- Infrared spectrum:Conforms
- Additional info:package usually contains small pouches of ammonium carbonate as preservative
- Solubility:(5 % in water) Passes test
- Residue after ignition:≤0.05 %
- Suitability:(for precip. of Fe, etc.) Passes test
Technische Daten
- Tipo di conf.:Bottiglia di plastica
- Conf:500 g



