Repair set
Repair set for Transferpettor™ 50 µl
Codice catalogo 612-5384
Nome del fornitore: Brand
Vai a
Product Details & Documents
Set includes 1 allen key, 1 piston rod with fitted PTFE Transferpettor™ seal (for capacities ≥ 20 µl), 1 calibration gauge, 1 screwdriver, 3 clamping discs, 1 fixing screw, 3 PTFE Transferpettor™ seals, 1 mounting block (for capacities ≥ 20 µl).
Documents
Brand Transferpettor
Letteratura sul prodotto
Specifiche
- Descrizione:Repair set for Transferpettor™ 50 µl
- Seq:04