Laboratory trays
652×530×140 mm
Codice catalogo BRND156648
Nome del fornitore: Brand
Vai a
Product Details & Documents
PP, white.
- Very good chemical resistance
- Very good thermal resistance
- Smooth surfaces for easy cleaning
With ribs on base. Highly versatile trays, ideal for a wide range of applications. The ribs on the base make these trays ideal for use as developing trays.
Specifiche
- L×P×A:652×530×140 mm