Funnel holders
2 funnels
Codice catalogo 241-0344
Nome del fornitore: Brand
Vai a
Product Details & Documents
PP, white.
To support 1 or 2 funnels with top outer-Ø of 50 to 120 mm. Easily mounted on supports rods with an outer-Ø 8 to 14 mm with a locking nut.
Specifiche
- Per:2 funnels