Funnel holders
Funnel holder for a funnel, 1 funnel
241-0343
: Brand
PP, white.
To support 1 or 2 funnels with top outer-Ø of 50 to 120 mm. Easily mounted on supports rods with an outer-Ø 8 to 14 mm with a locking nut.
- Per:1 funnel
- Descrizione:Funnel holder for a funnel