Imbuti
143 mm, 14 mm, 180 mm
Codice catalogo BRND148245
Nome del fornitore: Brand
Vai a
Product Details & Documents
PP, translucent.
- External flutings allow air to escape during filtration
- Excellent chemical resistance, autoclavable
- Suitable for use with standard filter papers
For rapid filtration.
Specifiche
- Ø esterno imbuto:180 mm
- Ø esterno stelo:14 mm
- Lunghezza asta:143 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



