D(-)-Luciferin
Bottiglia di vetro, 50 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
- CAS Number:2591-17-5
- Formula:C₁₁H₈N₂O₃S₂
- MDL Number:MFCD00042929
- Molecular Weight:280.3228 g/mol
- Melting Pt:200 °C
- Storage Temperature:Congelamento
- EINECS:219-981-3
Documents
ACRO33705
SDS
Specifications
Specification Test Results
- Appearance:Off-white to light yellow Powder
- Infrared spectrum:Conforms
- Water:≤2.0 % (K.F.)
- HPLC:≥97.5 %
- UV Lambda max:327 to 331 nm (c=1, CH3OH)
- Specific optical rotation:-32.0° to -36.0° (20°C, 589 nm) (c=1, DMF)
- Molar extinction coefficient:≥18200 l/(mol.cm) (327 to 331 nm, c=1, CH3OH)
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:50 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



