Orders LinkContactUsLinkComponent
HPLC columns, Aminex®
Guard cartridge for Aminex® fast carbohydrate analysis and HPX-87P column, Precartuccia, 4,60 mm, 30 mm
  BRAD125-0119
Marketsource
 :   BIO RAD
HPLC columns, Aminex®
  BRAD125-0119
 :  BIO RAD
Marketsource
HPLC columns, Aminex®
Guard cartridge for Aminex® fast carbohydrate analysis and HPX-87P column, Precartuccia, 4,60 mm, 30 mm
SFr. 692.00
1 * 2 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
  • Formato della colonna:
    Precartuccia
  • Ø int.:
    4,60 mm
  • Descrizione:
    Guard cartridge for Aminex® fast carbohydrate analysis and HPX-87P column
  • Lunghezza:
    30 mm
Accessories
  •  
    Descrizione
  • BRAD125-0131
    Standard cartridge holder for Ø×L 30×4,6 mm catridges
    1 * 1 ST - SFr. 978.00