Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Magnetic stirring bars, PTFE, triangular, BOLA

Magnetic stirring bars, PTFE, triangular, BOLA

35 mm, 10 mm, Recipienti di grandi dimensioni
Codice catalogo BOHLC357-15
Nome del fornitore:   Bohlender
Magnetic stirring bars, PTFE, triangular, BOLA
Codice catalogo BOHLC357-15
Nome del fornitore:  Bohlender
Magnetic stirring bars, PTFE, triangular, BOLA
Magnetic stirring bars, PTFE, triangular, BOLA

35 mm, 10 mm, Recipienti di grandi dimensioni

Price
SFr. 8.80
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Triangular shaped magnetic stirrer bars with magnetic core of Alnico 5 encapsulated in PTFE.

  • Chemical resistant

Temperature resistance: −200 to +250 °C.

Most commonly used for big vessels in which strong turbulence can be acheived with low speeds. These can also been useful for mixing of reagents which resist dissolving and for avoiding any residues at the bottom of the vessels.

Specifiche
  • Ø:
    10 mm
  • Per:
    Recipienti di grandi dimensioni
  • Lunghezza:
    35 mm
  • Seq:
    05