Sterilisation Boxes for Pipettes
Square base, 70×70×427 mm
Codice catalogo 612-1629
Nome del fornitore: BOCHEM
Vai a
Product Details & Documents
Stainless steel, with square base, autoclavable.
- Can be autoclaved at 121 °C and sterilised at 205 °C
Bottom and underside of the lid are coated in silicone to minimise the risk of damaging the pipettes during transport and sterilisation
Specifiche
- Descrizione:Square base
- L×P×A:70×70×427 mm