Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Sessole chimiche

Sessole chimiche

50 ml, 135 mm, 24 mm
Catalog # BOCH8816
Supplier:   BOCHEM
Sessole chimiche
Catalog # BOCH8816
Supplier:  BOCHEM
Sessole chimiche
Sessole chimiche

50 ml, 135 mm, 24 mm

Price
SFr. 4.10
Log in for availability
1 * 1 ST
Product Details & Documents

These scoops are made from aluminium.

Specifications
  • Ø:
    24 mm
  • Capacità:
    50 ml
  • Lunghezza:
    135 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.