Drums
400 mm
Catalog # BOCH8356
Supplier: BOCHEM
Jump to:
Product Details & Documents
Made of 18/10 stainless steel.
- With or without lid available
- With handle
- Very good temperature resistance
Specifications
- Ø est:400 mm
- Seq:18
Accessories
Product Information
- Catalog #DescrizioneAvailabilityPrice
- BOCH8341RCastor with 4 rubber guide rolls (1 lockable), for 25 L and 30 L containersLog in for availability1 * 1 ST - SFr. 326.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- BOCH8344RCastor with 4 rubber guide rolls (1 lockable), for 50 L containersLog in for availability1 * 1 ST - SFr. 348.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- BOCH8346RCastor with 4 rubber guide rolls (1 lockable), for 75 L containersLog in for availability1 * 1 ST - SFr. 375.00
Product Information
- Catalog #DescrizioneMaterialeAvailabilityPrice
- BOCH8349Morsetto di bloccaggio per contenitori, 3 pezzi, già montatoAcciaio inossidabile 18/10Log in for availability1 * 3 ST - SFr. 155.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- BOCH8348RCastor with 4 rubber guide rolls (1 lockable), for 150 L containersLog in for availability1 * 1 ST - SFr. 625.00
Product Information
- Catalog #DescrizioneAvailabilityPrice
- BOCH8347RCastor with 4 rubber guide rolls (1 lockable), for 100 L containersLog in for availability1 * 1 ST - SFr. 576.00