Cestelli
Coperchio per cestelli graduati 15 l, acciaio inox 18/10, Ø 310 mm
Catalog # 216-0233
Supplier: BOCHEM
Jump to:
Product Details & Documents
Buckets, stainless steel 18/10.
- Very good chemical resistance, very good thermal resistance
- High quality, very stable
- Matching lids available optionally
For use in laboratories and industry. Graduated with handle. Seamless handle tabs and smooth surfaces for fast, easy cleaning.
Specifications
- Descrizione:Coperchio per cestelli graduati 15 l, acciaio inox 18/10, Ø 310 mm
- Seq:11