Retort rods, with M10 thread
12 mm, 1250 mm, M10
Catalog # BOCH5114
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10.
- Rods for stand bases
- M10 thread
Specifications
- Ø:12 mm
- Filettatura:M10
- Lunghezza:1250 mm
- Seq:05
Stainless steel 18/10.