Stand bases
1,9 kg, 160×250×3 mm
Katalognr. BOCH5001
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
Stand bases made of stainless steel 18/10 with expanded, flat and heavy feet for stability.
- With M 10 threaded hole (off-centre)
- One adjustable foot
- Anti-slip rubber protection
Certifications: DIN 12892
Technische Daten
- L×P×A:160×250×3 mm
- Peso:1,9 kg