Stand bases
1,6 kg, 130×210×3 mm
Catalog # BOCH5000
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stand bases made of stainless steel 18/10 with expanded, flat and heavy feet for stability.
- With M 10 threaded hole (off-centre)
- One adjustable foot
- Anti-slip rubber protection
Certifications: DIN 12892
Specifications
- L×P×A:130×210×3 mm
- Peso:1,6 kg