Orders LinkContactUsLinkComponent
Forceps with guide pin
Forceps with guide pin
Bent, sharp, 115 mm
Codice catalogo 232-0018
Nome del fornitore:   BOCHEM
Forceps with guide pin
Codice catalogo 232-0018
Nome del fornitore:  BOCHEM
Forceps with guide pin
Forceps with guide pin
Bent, sharp, 115 mm
Price
42.40 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Stainless steel, with PTFE coating.
  • Bent, with guide pin
  • Smooth, coated surface to prevent contamination and facilitate cleaning
Specifiche
  • Tipo:
    Bent, sharp
  • Lunghezza:
    115 mm