Forceps with guide pin
Sharp, straight, 115 mm
Codice catalogo 232-0016
Nome del fornitore: BOCHEM
Vai a
Product Details & Documents
Stainless steel, PTFE coating.
- Anti-magnetic
- Very good chemical and thermal resistance
- Smooth surface prevents contamination and facilitates cleaning
Specifiche
- Tipo:Sharp, straight
- Lunghezza:115 mm