Forceps, precision
Straight, extra sharp, 120 mm
Codice catalogo 232-0009
Nome del fornitore: BOCHEM
Vai a
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic. Extremely precise forceps designed for a wide variety of uses in the laboratory, production etc.
- Extremely sharp tips without grooves
- Reverse action; closed normally, squeeze to open
Specifiche
- Tipo:Straight, extra sharp
- Lunghezza:120 mm