Orders LinkContactUsLinkComponent
Adapters, reduction, Quickfit®
Adapters, reduction, Quickfit®
24/29 NS
Codice catalogo 201-2375
Nome del fornitore:   SciLabware
Adapters, reduction, Quickfit®
Codice catalogo 201-2375
Nome del fornitore:  SciLabware
Adapters, reduction, Quickfit®
Adapters, reduction, Quickfit®
24/29 NS
Price
SFr. 105.00
Log in per disponibilità
1 * 2 ST
Product Details & Documents
PYREX®, borosilicate glass, Quickfit® joints, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.
Specifiche
  • Cono femmina:
    24/29 NS