Orders LinkContactUsLinkComponent
Adapters, receiver, vacuum, Quickfit®
19/26 NS, 19/26 NS, 140 mm
  201-8160
 :   SciLabware
Adapters, receiver, vacuum, Quickfit®
  201-8160
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
19/26 NS, 19/26 NS, 140 mm
SFr. 112.00
1 * 1 ST
PYREX®, borosilicate glass, bent.
  • Easy connection to flexible tubing via detachable plastic screw thread connector
Adapter with a side arm for connection to a vacuum supply.
  • Dimensione cono:
    19/26 NS
  • Cono femmina:
    19/26 NS
  • Lunghezza totale:
    140 mm