Non-return valves, SP Bel-Art
9,5 - 12,7 mm ID
Codice catalogo BELAH197170000
Nome del fornitore: Bel-Art Products, a Part of SP
Non-return valves, SP Bel-Art
Codice catalogo BELAH197170000
Nome del fornitore: Bel-Art Products, a Part of SP
Vai a
Product Details & Documents
PP valve, translucent with neoprene valve disk. Stops reverse flow of liquid when pressure is interrupted.
- Useful when connecting a vacuum line to a filter flask
- Recommended maximum pressure 20 psi. (1034 Torr)
- Autoclavable
Specifiche
- Dimensione del tubo:9,5 - 12,7 mm ID