Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
L(-)-Carnitine 99+%

L(-)-Carnitine 99+%

Bottiglia di vetro, 10 g
Catalog # ACRO241040100
Irritant
L(-)-Carnitine 99+%
Catalog # ACRO241040100
CAS Number:  541-15-1
L(-)-Carnitine 99+%
L(-)-Carnitine 99+%

Bottiglia di vetro, 10 g

CAS Number:  541-15-1
Price
SFr. 188.00
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
  • CAS Number:541-15-1
  • Formula:C₇H₁₅NO₃
  • MDL Number:MFCD00038747
  • Molecular Weight:161.20 g/mol
  • Melting Pt:208…212 °C (dec.)
  • Storage Temperature:Refrigerata
  • EINECS:208-768-0

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Solubility:
    (5 % in water) Clear colorless
  • Specific optical rotation:
    -29° to -32° (20°C, 589 nm) (c=1, H2O)
  • Titration with HClO4:
    ≥99.0 %

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    10 g

Specifications