Orders LinkContactUsLinkComponent
Unions
Union, LP 0,5 mm bore, complete, 1/16” tubing, PEEK
  554-1026
Unions
  554-1026
Unions
Union, LP 0,5 mm bore, complete, 1/16” tubing, PEEK
SFr. 60.20
1 * 1 ST
Unions for easy connection of low pressure solvent lines.
  • Low dead volume
  • Inert and biocompatible
  • Descrizione:
    Union, LP 0,5 mm bore, complete, 1/16” tubing
  • Materiale:
    PEEK
  • Seq:
    05