Unions
Union, LP 0,5 mm bore, complete, 1/16” tubing, PEEK
554-1026
Unions for easy connection of low pressure solvent lines.
- Low dead volume
- Inert and biocompatible
- Descrizione:Union, LP 0,5 mm bore, complete, 1/16” tubing
- Materiale:PEEK
- Seq:05