Unions
Union, LP 1,3 mm bore, body only, PEEK
JOURJR-065
Unions for easy connection of low pressure solvent lines.
- Low dead volume
- Inert and biocompatible
- Descrizione:Union, LP 1,3 mm bore, body only
- Materiale:PEEK
- Seq:04