Orders LinkContactUsLinkComponent
Unions
Union, LP 1,3 mm bore, body only, PEEK
  JOURJR-065
Unions
  JOURJR-065
Unions
Union, LP 1,3 mm bore, body only, PEEK
SFr. 32.80
1 * 1 ST
Unions for easy connection of low pressure solvent lines.
  • Low dead volume
  • Inert and biocompatible
  • Descrizione:
    Union, LP 1,3 mm bore, body only
  • Materiale:
    PEEK
  • Seq:
    04