Ancorette magnetiche, PTFE
Kit con ancorette magnetiche IKAFLON® 10, 15, 20, 25, 30, 40, 50, 80 e TRIKA® 25 e 40
441-2121
: IKA
- Very good chemical resistance
- Temperature resistant, suitable for high and low temperature applications
- Tipo:Kit con ancorette magnetiche IKAFLON® 10, 15, 20, 25, 30, 40, 50, 80 e TRIKA® 25 e 40