Ancorette magnetiche, PTFE
Curve, 155 mm, 27 mm
IKAA1129000
: IKA
- Very good chemical resistance
- Temperature resistant, suitable for high and low temperature applications
- Ø:27 mm
- Tipo:Curve
- Lunghezza:155 mm