Orders LinkContactUsLinkComponent
Ancorette magnetiche, PTFE
Ancorette magnetiche, PTFE
Curve, 155 mm, 27 mm
  IKAA1129000
 :   IKA
Ancorette magnetiche, PTFE
  IKAA1129000
 :  IKA
Ancorette magnetiche, PTFE
Ancorette magnetiche, PTFE
Curve, 155 mm, 27 mm
SFr. 172.00
1 * 1 ST
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications
  • Ø:
    27 mm
  • Tipo:
    Curve
  • Lunghezza:
    155 mm