Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Provette con bordo

100 mm, 12 mm, 0,8 mm
  HIRS9270107
 :   HIRSCHMANN
Provette con bordo
  HIRS9270107
Provette con bordo
Provette con bordo

100 mm, 12 mm, 0,8 mm

 
SFr. 14.60
   
1 * 100 ST
 

Soda-lime glass.

  • Good chemical resistance
  • Withstand moderate temperature change
  • Round bottom
 
  • Ø est:
    12 mm
  • Spessore parete:
    0,8 mm
  • Lunghezza:
    100 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.