Stopcocks for burettes, lateral
Burette di capacità 25 ml
Katalognr. HIRS8901170
Anbieter: HIRSCHMANN
Direkt zu
Product Details & Documents
Lateral burette stopcock, made of borosilicate glass, with glass key, clear glass.
- Excellent chemical resistance
- Excellent temperature resistance
Technische Daten
- Per:Burette di capacità 25 ml
- Seq:02