Dissolved oxygen bottles
Bombole per ossigeno secondo Winkler, 100-150 ml, 14/23
Catalog # 612-2794
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Oxygen bottles according to Winkler, with white inscription label.
- The adjusted capacity is individually marked on the bottle
- A unique serial number is engraved on the bottle and stopper
Specifications
- Capacità:100-150 ml
- Descrizione:Bombole per ossigeno secondo Winkler
- Tappo a molatura standard:14/23