Provette, con beccuccio
20 ml, 205 mm, 17 mm
Codice catalogo HIRS6400166
Nome del fornitore: HIRSCHMANN
Vai a
Product Details & Documents
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
- Graduated
Specifiche
- Ø est:17 mm
- Capacità:20 ml
- Lunghezza:205 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



