Provette, con beccuccio
20 ml, 205 mm, 17 mm
HIRS6400165
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
- Graduated
- Ø est:17 mm
- Capacità:20 ml
- Lunghezza:205 mm