Provette, con beccuccio
10 ml, 170 mm, 14 mm
HIRS6400160
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
- Graduated
- Ø est:14 mm
- Capacità:10 ml
- Lunghezza:170 mm