Provette per centrifuga
10 ml, ∼16×100 mm, graduated
HIRS6110101
Glass.
- Graduated or ungraduated
- Good chemical resistance
Long conical or short conical form.
- Ø×L:∼16×100 mm
- Tipo:graduated
- Capacità:10 ml
Glass.
Long conical or short conical form.