Automatic burettes, AS, Pellet pattern, lateral stopcocks and intermediate valve
25 ml, 0,05 ml, 500 mm, 0,03 ± ml
Catalog # HIRS355037060
Supplier: HIRSCHMANN
Automatic burettes, AS, Pellet pattern, lateral stopcocks and intermediate valve
Catalog # HIRS355037060
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Automatic burettes, Pellet pattern, class AS, brown glass, DURAN® with white graduation.
- Main point ring graduation
- Lateral PTFE stopcock with intermediate valve
Certifications: Compliance with DIN EN ISO 385.
Specifications
- Capacità:25 ml
- Divisione:0,05 ml
- Tolleranza:0,03 ± ml
- Lunghezza scala:500 mm