Instrument dishes
200×100×100 mm
Catalog # 216-1408
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Thick-walled glass.
- Can be sterilised in hot air up to 180 °C
- Robust
- Stainless steel lids are available separately
Multipurpose dishes with vertical walls for a variety of applications in the laboratory, field use and industry.
Specifications
- Seq:04
- L×P×A:200×100×100 mm