Instrument dishes
120×60×40 mm
Codice catalogo 216-1401
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
Thick-walled glass.
- Can be sterilised in hot air up to 180 °C
- Robust
- Stainless steel lids are available separately
Multipurpose dishes with vertical walls for a variety of applications in the laboratory, field use and industry.
Specifiche
- Seq:01
- L×P×A:120×60×40 mm