Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Test Tubes, round bottom, with spout

Test Tubes, round bottom, with spout

20 ml, 200 mm, 15 mm
Codice catalogo 212-1606
Nome del fornitore:   GLASWARENFABRIK KARL HECHT
Test Tubes, round bottom, with spout
Codice catalogo 212-1606
Nome del fornitore:  GLASWARENFABRIK KARL HECHT
Test Tubes, round bottom, with spout
Test Tubes, round bottom, with spout

20 ml, 200 mm, 15 mm

Price
SFr. 6.70
Log in per disponibilità
1 * 1 ST
Product Details & Documents

AR® (soda glass).

  • Fused white graduation
  • Good chemical and physical properties
  • Round bottom

Suitable for use in applications that do not involve high thermal stresses, especially for bacteriological applications.

Specifiche
  • Ø est:
    15 mm
  • Capacità:
    20 ml
  • Lunghezza:
    200 mm