Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Knife spatulas

Knife spatulas

250 mm, 150 mm
Catalog # HAMMHWN253-25
Supplier:   HAMMACHER KARL
Knife spatulas
Catalog # HAMMHWN253-25
Supplier:  HAMMACHER KARL
Knife spatulas
Knife spatulas

250 mm, 150 mm

Price
SFr. 69.60
Log in for availability
1 * 1 ST
Product Details & Documents

Knife spatulas with wooden handle and blade made of stainless WIRONIT steel.

Specifications
  • Lunghezza:
    250 mm
  • Seq:
    04
  • Lunghezza lama:
    150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.