Scalpels
155 - 160 mm, 40 mm, Broad blade
Codice catalogo 233-0094
Nome del fornitore: HAMMACHER KARL
Vai a
Product Details & Documents
These scalpels are made from Wironit steel (18/12 stainless steel with 18% chrome and from 10 to 12% nickel).
- Resistant to corrosion and anti-magnetic
- With fixed blade and metal handle
Specifiche
- Descrizione:Broad blade
- Lunghezza:155 - 160 mm
- Seq:03
- Lama:40 mm