Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Preparation knives

Preparation knife, very delicate, oval blade, 20 mm, 120 mm
  233-0090
Preparation knives
  233-0090
Preparation knives
Preparation knives

Preparation knife, very delicate, oval blade, 20 mm, 120 mm

 
SFr. 109.00
   
1 * 1 ST
 

WIRONIT steel.

 
  • Tipo:
    Preparation knife, very delicate, oval blade
  • Seq:
    03
  • Lunghezza totale:
    120 mm
  • Lama:
    20 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.