Preparation knives
Preparation knife, very delicate, 20 mm, 120 mm
233-0087
WIRONIT steel.
- Tipo:Preparation knife, very delicate
- Seq:01
- Lunghezza totale:120 mm
- Lama:20 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



