Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Cation exchange bulk packing material

PRP-X400 bulk resin, 30 - 50 µm, -, 1 g
Catalog # 553-0014
Supplier:   HAMILTON BONADUZ
Cation exchange bulk packing material
Catalog # 553-0014
Supplier:  HAMILTON BONADUZ
CAS Number:  
Cation exchange bulk packing material
Cation exchange bulk packing material

PRP-X400 bulk resin, 30 - 50 µm, -, 1 g

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents

Polymer bulk packing material for cation exchange applications.

Chromatographic capacity and efficiency tests are conducted on both bulk and packed column resins to ensure product integrity.

Specifications

Specifications

  • Dimensioni particelle:
    30 - 50 µm
  • Conf:
    1 g
  • Dimensioni pori:
    -
  • Descrizione:
    PRP-X400 bulk resin

Specifications