Cation exchange bulk packing material
PRP-X400 bulk resin, 12 - 20 µm, -, 1 g
Codice catalogo 553-0013
Nome del fornitore: HAMILTON BONADUZ
Cation exchange bulk packing material
Codice catalogo 553-0013
Nome del fornitore: HAMILTON BONADUZ
Numero CAS:
Vai a
Product Details & Documents
Polymer bulk packing material for cation exchange applications.
Chromatographic capacity and efficiency tests are conducted on both bulk and packed column resins to ensure product integrity.
Specifiche
Specifiche
- Dimensioni particelle:12 - 20 µm
- Conf:1 g
- Dimensioni pori:-
- Descrizione:PRP-X400 bulk resin
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



